Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Kenpaullone 98%

Supplier: AMBEED, INC
Warning

A196851-10MG A196851-1MG A196851-50MG A196851-5MG
76800-204EA 39 USD InStock
76800-204 76800-206 76800-208 76800-210
Kenpaullone 98%
Kenpaullone
Precaución: for Research Use Only


Formula: C₁₆H₁₁BrN₂O
MDL Number: MFCD02683595
CAS Number: 142273-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty