Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-methyl-2-phenylpropanoate 97%

Supplier: AMBEED, INC
Warning

A197216-100G A197216-25G A197216-500G A197216-5G
76800-216EA 100 USD InStock
76800-216 76800-218 76800-220 76800-222 76800-224
Ethyl-2-methyl-2-phenylpropanoate 97%
Ethyl-2-methyl-2-phenylpropanoate
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₂
MDL Number: MFCD03844687
CAS Number: 2901-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty