Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1H-pyrazole-4-carboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A202559-100G A202559-1G A202559-25G A202559-5G
76800-398EA 216 USD InStock
76800-398 76800-400 76800-402 76800-404
Methyl 1H-pyrazole-4-carboxylate hydrochloride 95%
Methyl 1H-pyrazole-4-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₅H₇ClN₂O₂
MDL Number: MFCD08275052
CAS Number: 181997-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty