Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium ferulate 99+%

Supplier: AMBEED, INC
Warning

A203361-100G A203361-100MG A203361-1MG A203361-5G A203361-250MG A203361-25G
76800-442EA 117 USD InStock
76800-442 76800-444 76800-446 77567-944 77600-662 77667-698
Sodium ferulate 99+%
Sodium ferulate
Precaución: For research use only.


Formula: C₁₀H₉NaO₄
MDL Number: MFCD00075727
CAS Number: 24276-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty