Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl maleate 95%

Supplier: AMBEED, INC
Warning

A203981-100G A203981-25G A203981-500G
76800-454EA 12 USD InStock
76800-454 76800-456 76800-458
Dimethyl maleate 95%
Dimethyl maleate
Precaución: for Research Use Only


Formula: C₆H₈O₄
MDL Number: MFCD00008459
CAS Number: 624-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty