Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fumaric acid xpotassium salt 95%

Supplier: AMBEED, INC
Warning

A209158-1G A209158-250MG A209158-25G A209158-5G
76800-566EA 6 USD InStock
76800-566 76800-568 76800-570 76800-572
Fumaric acid xpotassium salt 95%
Fumaric acid xpotassium salt
Precaución: for Research Use Only


Formula: C₄H₄O₄·xK
MDL Number: MFCD00039096
CAS Number: 7704-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty