Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl myristate 97%

Supplier: AMBEED, INC
Warning

A209680-100G A209680-10G A209680-25G
76800-604EA 10 USD InStock
76800-604 76800-606 76800-608
Ethyl myristate 97%
Ethyl myristate
Precaución: for Research Use Only


Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
MDL Number: MFCD00008984
CAS Number: 124-06-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty