Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl pimelate 98%

Supplier: AMBEED, INC
Warning

A210172-100G A210172-10G A210172-25G
76800-616EA 41 USD InStock
76800-616 76800-618 76800-620
Dimethyl pimelate 98%
Dimethyl pimelate
Precaución: for Research Use Only


Formula: C₉H₁₆O₄
MDL Number: MFCD00008470
CAS Number: 1732-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty