Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Maltol propionate 97%

Supplier: AMBEED, INC
Warning

A210302-100G A210302-10G A210302-25G A210302-5G
76800-622EA 245 USD InStock
76800-622 76800-624 76800-626 76800-628
Maltol propionate 97%
Maltol propionate
Precaución: For research use only.


Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00036605
CAS Number: 68555-63-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty