Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-D-Glu(Obzl)-Obzl P-tosylate 98%

Supplier: AMBEED, INC
Warning

A213649-100G A213649-25G A213649-5G
76800-706EA 37 USD InStock
76800-706 76800-708 76800-710
H-D-Glu(Obzl)-Obzl P-tosylate 98%
H-D-Glu(Obzl)-Obzl P-tosylate
Precaución: for Research Use Only


Formula: C₂₆H₂₉NO₇S
MW: 499.578 g/mol
MDL Number: MFCD00153435
CAS Number: 19898-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty