Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6-phenylpyridazine 96%

Supplier: AMBEED, INC
Warning

A213954-10G A213954-1G A213954-250MG A213954-5G
76800-716EA 228 USD InStock
76800-716 76800-718 76800-720 76800-722
3-Chloro-6-phenylpyridazine 96%
3-Chloro-6-phenylpyridazine
Precaución: for Research Use Only


Formula: C₁₀H₇ClN₂
MDL Number: MFCD00075253
CAS Number: 20375-65-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty