Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl toluene-4-sulphonate 98%

Supplier: AMBEED, INC
Warning

A216764-10G A216764-500G A216764-5G A216764-25G A216764-100G
76800-782EA 7 USD InStock
76800-782 76800-784 76800-786 77303-436 77667-818
Ethyl toluene-4-sulphonate 98%
Ethyl toluene-4-sulphonate
Precaución: for Research Use Only


Formula: C₉H₁₂O₃S
MDL Number: MFCD00009100
CAS Number: 80-40-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty