Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-tert-Butylcyclohexanecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A217854-1G A217854-250MG A217854-5G
76800-804EA 35 USD InStock
76800-804 76800-806 76800-808
trans-4-tert-Butylcyclohexanecarboxylic acid 97%
trans-4-tert-Butylcyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₂₀O₂
MDL Number: MFCD00466237
CAS Number: 943-29-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty