Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl (trimethylsilyl)acetate 96%

Supplier: AMBEED, INC
Danger

A219334-10G A219334-1G A219334-25G A219334-5G
76800-848EA 11 USD InStock
76800-848 76800-850 76800-852 76800-854
Ethyl (trimethylsilyl)acetate 96%
Ethyl (trimethylsilyl)acetate
Precaución: for Research Use Only


Formula: C₇H₁₆O₂Si
MW: 160.29 g/mol
MDL Number: MFCD00009172
CAS Number: 4071-88-9
UN: 3272
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty