Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Epmedin C 98%

Supplier: AMBEED, INC
Warning

A228815-10MG A228815-1MG A228815-50MG A228815-5MG
76801-036EA 47 USD InStock
76801-036 76801-038 76801-040 76801-042
Epmedin C 98%
Epmedin C
Precaución: for Research Use Only


Formula: C₃₉H₅₀O₁₉
MDL Number: MFCD04039820
CAS Number: 110642-44-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty