Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-fluoroacetoacetate 95%

Supplier: AMBEED, INC
Warning

A230103-100G A230103-25G A230103-5G
76801-076EA 116 USD InStock
76801-076 76801-078 76801-080
Ethyl-2-fluoroacetoacetate 95%
Ethyl-2-fluoroacetoacetate
Precaución: for Research Use Only


Formula: C₆H₉FO₃
MDL Number: MFCD00215830
CAS Number: 1522-41-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty