Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Nα-methyl-O-benzyl-L-threonine 98%

Supplier: AMBEED, INC
Warning

A230737-1G A230737-5G
76801-084EA 50 USD InStock
76801-084 76801-086
Fmoc-Nα-methyl-O-benzyl-L-threonine 98%
Fmoc-Nalpha-methyl-O-benzyl-L-threonine
Precaución: for Research Use Only


Formula: C₂₇H₂₇NO₅
MDL Number: MFCD00235882
CAS Number: 198561-81-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty