Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Efaproxiral sodium ≥98%

Supplier: AMBEED, INC
Warning

A230819-10MG A230819-1MG A230819-50MG
76801-088EA 8 USD InStock
76801-088 76801-090 76801-092
Efaproxiral sodium ≥98%
Efaproxiral sodium
Precaución: for Research Use Only


Formula: C₂₀H₂₂NNaO₄
MDL Number: MFCD02093864
CAS Number: 170787-99-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty