Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-benzofuran-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A235291-1G A235291-25G A235291-5G
76801-168EA 17 USD InStock
76801-168 76801-170 76801-172
Ethyl-1-benzofuran-2-carboxylate 97%
Ethyl-1-benzofuran-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₀O₃
MDL Number: MFCD03094950
CAS Number: 3199-61-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty