Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A240087-100G A240087-10G A240087-25G A240087-5G
76801-306EA 279 USD InStock
76801-306 76801-308 76801-310 76801-312
Ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate 97%
Ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₁NO₄
MDL Number: MFCD00205625
CAS Number: 10350-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty