Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mefenpyr-diethyl 95%

Supplier: AMBEED, INC
Warning

A240975-1000G A240975-100G A240975-10G A240975-25G A240975-500G A240975-5G
76801-330EA 87 USD InStock
76801-330 76801-332 76801-334 76801-336 76801-338 76801-340 76801-342
Mefenpyr-diethyl 95%
Mefenpyr-diethyl
Precaución: for Research Use Only


Formula: C₁₆H₁₈Cl₂N₂O₄
MW: 373.23 g/mol
MDL Number: MFCD09753375
CAS Number: 135590-91-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty