Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxyacetone 97% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: ≤500 ppm Stabilised with sodium carbonate (Na₂CO₃)

A244029-25G A244029-100G
76801-394EA 9 USD InStock
76801-394 77668-000
Hydroxyacetone 97% stabilised
Hydroxyacetone
Precaución: for Research Use Only


Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 145
Melting Pt: -17
MDL Number: MFCD00004669
CAS Number: 116-09-6
UN: 1224
ADR: 3 Ⅲ
Merck Index: 13,00066
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty