Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl butylmalonate 99%

Supplier: AMBEED, INC
Warning

A244917-100G A244917-25G A244917-500G
76801-408EA 25 USD InStock
76801-408 76801-410 76801-412
Diethyl butylmalonate 99%
Diethyl butylmalonate
Precaución: for Research Use Only


Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 235
MDL Number: MFCD00009170
CAS Number: 133-08-4
Merck Index: 13,01575
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty