Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Folinic acid calcium salt pentahydrate 98%

Supplier: AMBEED, INC
Warning

A246079-100MG A246079-1G A246079-250MG A246079-5G
76801-436EA 5 USD InStock
76801-436 76801-438 76801-440 76801-442
Folinic acid calcium salt pentahydrate 98%
Folinic acid calcium salt pentahydrate
Precaución: for Research Use Only


Formula: C₂₀H₃₁CaN₇O₁₂
MDL Number: MFCD00149465
CAS Number: 6035-45-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty