Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2,5-Diaminopentanoic acid acetic acid salt 95%

Supplier: AMBEED, INC
Warning

A247290-100G A247290-10G A247290-25G
76801-478EA 42 USD InStock
76801-478 76801-480 76801-482
(S)-2,5-Diaminopentanoic acid acetic acid salt 95%
(S)-2,5-Diaminopentanoic acid acetic acid salt
Precaución: for Research Use Only


Formula: C₇H₁₆N₂O₄
MDL Number: MFCD00077444
CAS Number: 60259-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty