Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminobenzene sulphonic acid 98%

Supplier: AMBEED, INC
Warning

A250699-1000G A250699-100G A250699-1G A250699-500G
76801-552EA 45 USD InStock
76801-552 76801-554 76801-556 76801-558
3-Aminobenzene sulphonic acid 98%
3-Aminobenzene sulphonic acid
Precaución: for Research Use Only


Formula: C₆H₇NO₃S
MW: 173.19 g/mol
MDL Number: MFCD00065345
CAS Number: 121-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty