Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenyl-4H-chromen-4-one 40%

Supplier: AMBEED, INC
Warning

A253709-100G A253709-25G
76801-604EA 24 USD InStock
76801-604 77668-068
3-Phenyl-4H-chromen-4-one 40%
3-Phenyl-4H-chromen-4-one
Precaución: for Research Use Only


Formula: C₁₅H₁₀O₂
MDL Number: MFCD00100851
CAS Number: 574-12-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty