Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lapatinib 98%

Supplier: AMBEED, INC
Warning

A254504-100MG A254504-1G A254504-1MG A254504-250MG A254504-25G A254504-50MG A254504-5G
76801-624EA 37 USD InStock
76801-624 76801-626 76801-628 76801-630 76801-632 76801-634 76801-636
Lapatinib 98%
Lapatinib
Precaución: for Research Use Only


Formula: C₂₉H₂₆ClFN₄O₄S
MDL Number: MFCD09264194
CAS Number: 231277-92-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty