Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Magnesium 2-hydroxypropanoate trihydrate 98%

Supplier: AMBEED, INC
Warning

A258763-1000G A258763-100G A258763-25G A258763-500G
76801-734EA 182 USD InStock
76801-734 76801-736 76801-738 76801-740
Magnesium 2-hydroxypropanoate trihydrate 98%
Magnesium 2-hydroxypropanoate trihydrate
Precaución: for Research Use Only


Formula: C₆H₁₆MgO₉
MDL Number: MFCD00156534
CAS Number: 18917-96-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty