Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indol-3-ylacetonitrile 98%

Supplier: AMBEED, INC
Danger

A258813-10G A258813-5G
76801-742EA 12 USD InStock
76801-742 76801-744
Indol-3-ylacetonitrile 98%
Indol-3-ylacetonitrile
Precaución: for Research Use Only


Formula: C₁₀H₈N₂
MW: 156.19 g/mol
MDL Number: MFCD00005628
CAS Number: 771-51-7
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty