Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Aspartic acid sodium 98%

Supplier: AMBEED, INC
Warning

A261187-1000G A261187-100G A261187-1G A261187-25G A261187-500G A261187-5G
76801-784EA 886 USD InStock
76801-784 76801-786 76801-788 76801-790 76801-792 76801-794
L-Aspartic acid sodium 98%
L-Aspartic acid sodium
Precaución: for Research Use Only


Formula: C₄H₅NNa₂O₄
MW: 177.06 g/mol
MDL Number: MFCD00082327
CAS Number: 5598-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty