Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ε-Hexalactone 97%

Supplier: AMBEED, INC
Warning

A262678-1000G A262678-100G A262678-25G A262678-500G
76801-820EA 47 USD InStock
76801-820 76801-822 76801-824 76801-826
ε-Hexalactone 97%
ε-Hexalactone
Precaución: for Research Use Only


Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 225
Melting Pt: -1
MDL Number: MFCD00003267
CAS Number: 502-44-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty