Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium (2R,3R)-2,3-dihydroxysuccinate 97%

Supplier: AMBEED, INC
Warning

A265767-100G A265767-10G A265767-25G A265767-500G A265767-5G
76801-880EA 46 USD InStock
76801-880 76801-882 76801-884 76801-886 76801-888
(R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium (2R,3R)-2,3-dihydroxysuccinate 97%
(R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium (2R,3R)-2,3-dihydroxysuccinate
Precaución: for Research Use Only


Formula: C₁₈H₃₆N₂O₁₂
MDL Number: MFCD04974128
CAS Number: 36687-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty