Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1-methyl-1H-pyrrole-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A272412-100G A272412-10G A272412-1G A272412-25G A272412-5G
76801-996EA 384 USD InStock
76801-996 76801-998 76802-000 76802-002 76802-004
Methyl-1-methyl-1H-pyrrole-2-carboxylate 98%
Methyl-1-methyl-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only


Formula: C₇H₉NO₂
MDL Number: MFCD00052747
CAS Number: 37619-24-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty