Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-amino-5-methylbenzoate 95%

Supplier: AMBEED, INC
Warning

A274421-100G A274421-10G A274421-25G A274421-5G
76802-072EA 70 USD InStock
76802-072 76802-074 76802-076 76802-078
Ethyl-2-amino-5-methylbenzoate 95%
Ethyl-2-amino-5-methylbenzoate


Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00115547
CAS Number: 58677-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty