Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-methylbenzoylformate 95%

Supplier: AMBEED, INC
Warning

A274675-1G A274675-25G A274675-5G
76802-082EA 23 USD InStock
76802-082 76802-084 76802-086
Ethyl-4-methylbenzoylformate 95%
Ethyl 4-methylbenzoylformate
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₃
MDL Number: MFCD01311169
CAS Number: 5524-56-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty