Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine 98%

Supplier: AMBEED, INC
Warning

A274856-100MG A274856-1G A274856-250MG
76802-092EA 48 USD InStock
76802-092 76802-094 76802-096
4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine 98%
4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine
Precaución: for Research Use Only


Formula: C₁₁H₁₀ClN₅
MW: 247.69 g/mol
MDL Number: MFCD00059781
CAS Number: 33006-91-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty