Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-bromo-3,3,3-trifluoropropanoate 97%

Supplier: AMBEED, INC
Danger

A275897-100MG A275897-1G A275897-250MG
76802-108EA 79 USD InStock
76802-108 76802-110 76802-112
Methyl-2-bromo-3,3,3-trifluoropropanoate 97%
Methyl-2-bromo-3,3,3-trifluoropropanoate
Precaución: for Research Use Only


Formula: C₄H₄BrF₃O₂
MDL Number: MFCD04038319
CAS Number: 113816-36-7
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty