Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-D-Ser(tBu)-OH 96%

Supplier: AMBEED, INC
Warning

A278634-10G A278634-1G A278634-250MG A278634-5G A278634-25G
76802-150EA 25 USD InStock
76802-150 76802-152 76802-154 76802-156 77668-228
H-D-Ser(tBu)-OH 96%
H-D-Ser(tBu)-OH
Precaución: for Research Use Only


Formula: C₇H₁₅NO₃
MDL Number: MFCD00077107
CAS Number: 18783-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty