Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-benzoylpropionate 98%

Supplier: AMBEED, INC
Warning

A289578-10G A289578-1G A289578-5G
76802-358EA 15 USD InStock
76802-358 76802-360 76802-362
Methyl-3-benzoylpropionate 98%
Methyl-3-benzoylpropionate
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
MDL Number: MFCD00014906
CAS Number: 25333-24-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty