Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl sulphone 99%

Supplier: AMBEED, INC
Warning

A290644-100G A290644-25G A290644-500G
76802-378EA 6 USD InStock
76802-378 76802-380 76802-382
Dimethyl sulphone 99%
Dimethyl sulphone
Precaución: for Research Use Only


Formula: C₂H₆O₂S
MW: 94.13 g/mol
Boiling Pt: 238
MDL Number: MFCD00007566
CAS Number: 67-71-0
Merck Index: 13,03284
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty