Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-pyridineboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A292279-100MG A292279-10G A292279-1G A292279-250MG A292279-25G
76802-394EA 5 USD InStock
76802-394 76802-396 76802-398 76802-400 76802-402
5-Bromo-3-pyridineboronic acid pinacol ester 97%
5-Bromo-3-pyridineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₅BBrNO₂
MW: 283.96 g/mol
MDL Number: MFCD06798248
CAS Number: 452972-13-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty