Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Dimethylpiperidinium chloride 97%

Supplier: AMBEED, INC
Warning

A293658-100G A293658-10G A293658-25G
76802-422EA 50 USD InStock
76802-422 76802-424 76802-426 76802-428
1,1-Dimethylpiperidinium chloride 97%
1,1-Dimethylpiperidinium chloride
Precaución: for Research Use Only


Formula: C₇H₁₆ClN
MDL Number: MFCD00055543
CAS Number: 24307-26-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty