Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethylphosphonoacetic acid 98%

Supplier: AMBEED, INC
Warning

A295949-100G A295949-1G A295949-25G A295949-500G A295949-5G
76802-446EA 47 USD InStock
76802-446 76802-448 76802-450 76802-452 76802-454
Diethylphosphonoacetic acid 98%
Diethylphosphonoacetic acid
Precaución: for Research Use Only


Formula: C₆H₁₃O₅P
MDL Number: MFCD00192032
CAS Number: 3095-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty