Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl succinate 98%

Supplier: AMBEED, INC
Warning

A297183-1000G A297183-100G A297183-25G A297183-500G
76802-486EA 30 USD InStock
76802-486 76802-488 76802-490 76802-492
Diethyl succinate 98%
Diethyl succinate
Precaución: for Research Use Only


Formula: C₈H₁₄O₄
MDL Number: MFCD00009208
CAS Number: 123-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty