Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-chloro-6-phenylpyrimidine 98%

Supplier: AMBEED, INC
Warning

A299291-100MG A299291-250MG A299291-5G
76802-510EA 27 USD InStock
76802-510 76802-512 76802-514
2-Amino-4-chloro-6-phenylpyrimidine 98%
2-Amino-4-chloro-6-phenylpyrimidine
Precaución: for Research Use Only


Formula: C₁₀H₈ClN₃
MW: 205.64 g/mol
MDL Number: MFCD00030784
CAS Number: 36314-97-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty