Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanosine 5'-triphosphate trisodium salt 98%

Supplier: AMBEED, INC
Warning

A300448-100MG A300448-1G A300448-250MG
76802-518EA 62 USD InStock
76802-518 76802-520 76802-522
Guanosine 5'-triphosphate trisodium salt 98%
Guanosine 5'-triphosphate trisodium salt
Precaución: for Research Use Only


Formula: C₁₀H₁₃N₅Na₃O₁₄P₃
MDL Number: MFCD00077781
CAS Number: 36051-31-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty