Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Ethyl-1-methylazepan-3-yl)phenol hydrochloride 98%

Supplier: AMBEED, INC
Warning

A302225-100MG A302225-10MG A302225-1MG A302225-25MG A302225-50MG
76802-544EA 161 USD InStock
76802-544 76802-546 76802-548 76802-550 76802-552
3-(3-Ethyl-1-methylazepan-3-yl)phenol hydrochloride 98%
3-(3-Ethyl-1-methylazepan-3-yl)phenol hydrochloride
Precaución: for Research Use Only


Formula: C₁₅H₂₄ClNO
MDL Number: MFCD00941476
CAS Number: 59263-76-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty