Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium bis(oxalato)borate 95%

Supplier: AMBEED, INC
Warning

A304321-25G A304321-500G A304321-5G
76802-590EA 32 USD InStock
76802-590 76802-592 77550-834
Lithium bis(oxalato)borate 95%
Lithium Bis(oxalato)borate
Precaución: for Research Use Only


Formula: C₄BLiO₈
MW: 193.79 g/mol
MDL Number: MFCD07776904
CAS Number: 244761-29-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty