Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

h-d-Tyr-otbu 97%

Supplier: AMBEED, INC
Warning

A306741-10G A306741-1G A306741-250MG A306741-5G
76802-668EA 69 USD InStock
76802-668 76802-670 76802-672 76802-674
h-d-Tyr-otbu 97%
h-d-Tyr-otbu
Precaución: for Research Use Only


Formula: C₁₃H₁₉NO₃
MW: 237.299 g/mol
MDL Number: MFCD00191035
CAS Number: 87553-74-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty